Assay Details
| Gene Symbol: Nmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290368.1 | NP_001277297.1 | 3 - 4 | 370 | 95 | |
| NM_008708.2 | NP_032734.1 | 3 - 4 | 370 | 127 | ||
| GenBank mRNA | AF043327.1 | - | 3 - 4 | 294 | 127 | |
| AK134521.1 | - | 3 - 4 | 334 | 127 | ||
| AK161490.1 | - | 3 - 4 | 370 | 95 | ||
| AK168171.1 | - | 3 - 4 | 338 | 127 | ||
| AK168190.1 | - | 3 - 4 | 338 | 127 | ||
More Information
Set Membership: |
|

Nmt2
Genomic Map