Assay Details
| Gene Symbol: Nmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008708.2 | NP_032734.1 | 4 - 5 | 427 | 93 | |
| GenBank mRNA | AF043327.1 | - | 4 - 5 | 351 | 93 | |
| AK134521.1 | - | 4 - 5 | 391 | 93 | ||
| AK168171.1 | - | 4 - 5 | 395 | 93 | ||
| AK168190.1 | - | 4 - 5 | 395 | 93 | ||
More Information
Set Membership: |
|

Nmt2
Genomic Map