Assay Details
| Gene Symbol: Mcm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027290.3 | NP_081566.2 | 15 - 16 | 2329 | 80 | |
| GenBank mRNA | AK030780.1 | - | 15 - 16 | 2274 | 80 | |
| BC023236.1 | - | 7 - 8 | 1191 | 80 | ||
| BC039663.1 | - | 1 - 2 | 178 | 80 | ||
| BC064314.1 | - | 15 - 16 | 2692 | 80 | ||
| BC094576.1 | - | 15 - 16 | 2329 | 6853824 | 80 | |
| BC120687.1 | - | 14 - 15 | 2238 | 40129610 | 80 | |
| BC120689.1 | - | 14 - 15 | 2238 | 40129612 | 80 | |
More Information
Set Membership: |
|

Mcm10
Genomic Map