Assay Details
| Gene Symbol: Phyh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010726.2 | NP_034856.1 | 3 - 4 | 318 | 121 | |
| GenBank mRNA | AF023463.1 | - | 3 - 4 | 283 | 121 | |
| AK146753.1 | - | 3 - 4 | 303 | 121 | ||
| AK151372.1 | - | 3 - 4 | 266 | 119 | ||
| AK164495.1 | - | 3 - 4 | 269 | 121 | ||
| AK165412.1 | - | 3 - 4 | 262 | 121 | ||
| AK168648.1 | - | 3 - 4 | 288 | 121 | ||
| AK169213.1 | - | 3 - 4 | 260 | 121 | ||
| BC002018.1 | - | 3 - 4 | 295 | 3489939 | 121 | |
| BY139974.1 | - | 3 - 4 | 318 | 121 | ||
| D88670.1 | - | 3 - 4 | 278 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Phyh
Genomic Map