Assay Details
| Gene Symbol: Phyh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010726.2 | NP_034856.1 | 6 - 7 | 750 | 69 | |
| GenBank mRNA | AF023463.1 | - | 6 - 7 | 715 | 69 | |
| AK146753.1 | - | 6 - 7 | 735 | 69 | ||
| AK151372.1 | - | 6 - 7 | 698 | 69 | ||
| AK164495.1 | - | 6 - 7 | 701 | 69 | ||
| AK165412.1 | - | 6 - 7 | 694 | 69 | ||
| AK168648.1 | - | 6 - 7 | 720 | 69 | ||
| AK169213.1 | - | 6 - 7 | 692 | 69 | ||
| BC002018.1 | - | 6 - 7 | 727 | 3489939 | 69 | |
| D88670.1 | - | 6 - 7 | 710 | 69 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Phyh
Genomic Map