Assay Details
| Gene Symbol: Pbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290576.1 | NP_001277505.1 | 3 - 4 | 641 | 97 | |
| NM_016768.2 | NP_058048.1 | 3 - 4 | 641 | 97 | ||
| GenBank mRNA | AF020199.1 | - | 3 - 4 | 632 | 97 | |
| AF020200.1 | - | 3 - 4 | 634 | 97 | ||
| AK144363.1 | - | 3 - 4 | 558 | 97 | ||
| BC057879.1 | - | 3 - 4 | 532 | 5348692 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pbx3
Genomic Map