Assay Details
| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009261.3 | NP_033287.2 | 7 - 8 | 1003 | 64 | |
| GenBank mRNA | BC014710.1 | - | 8 - 9 | 1087 | 4502840 | 64 |
| X84692.1 | - | 940 | 64 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Strbp
Genomic Map