Assay Details
| Gene Symbol: Phf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028716.4 | NP_082992.1 | 7 - 8 | 748 | 65 | |
| GenBank mRNA | AK014030.1 | - | 6 - 7 | 1168 | 65 | |
| BC027776.1 | - | 7 - 8 | 743 | 5352251 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phf19
Genomic Map