Assay Details
| Gene Symbol: Ggta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF297615.1 | - | 1 - 2 | 188 | 79 | |
| AK004753.1 | - | 1 - 2 | 252 | 79 | ||
| AK088605.1 | - | 1 - 2 | 53 | 79 | ||
| AK159087.1 | - | 2 - 3 | 298 | 79 | ||
| BC006810.1 | - | 1 - 2 | 44 | 3154188 | 79 | |
| M26925.1 | - | 1 - 2 | 69 | 79 | ||
| M85153.1 | - | 1 - 2 | 132 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF297615.1, AK004753.1, AK088605.1, AK159087.1, BC006810.1, M26925.1, M85153.1 transcripts.
Set Membership: |
|

Ggta1
Genomic Map