Assay Details
| Gene Symbol: Hspa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163434.1 | NP_001156906.1 | 2 - 3 | 274 | 62 | |
| NM_022310.3 | NP_071705.3 | 1 - 2 | 356 | 62 | ||
| GenBank mRNA | AJ002387.1 | - | 1 - 2 | 135 | 62 | |
| AK004578.1 | - | 1 - 2 | 351 | 62 | ||
| AK076079.1 | - | 2 - 3 | 270 | 62 | ||
| AK077463.1 | - | 2 - 3 | 274 | 62 | ||
| AK144650.1 | - | 2 - 3 | 270 | 62 | ||
| AK144742.1 | - | 2 - 3 | 270 | 62 | ||
| AK146647.1 | - | 2 - 3 | 273 | 62 | ||
| AK148539.1 | - | 2 - 3 | 273 | 62 | ||
| AK149447.1 | - | 2 - 3 | 273 | 62 | ||
| AK151244.1 | - | 1 - 2 | 222 | 62 | ||
| AK151647.1 | - | 2 - 3 | 272 | 62 | ||
| AK151806.1 | - | 2 - 3 | 273 | 62 | ||
| AK152020.1 | - | 2 - 3 | 270 | 62 | ||
| AK152521.1 | - | 1 - 2 | 353 | 62 | ||
| AK159719.1 | - | 2 - 3 | 270 | 62 | ||
| AK160821.1 | - | 274 | 62 | |||
| AK166207.1 | - | 2 - 3 | 270 | 62 | ||
| AK166739.1 | - | 2 - 3 | 270 | 62 | ||
| AK167011.1 | - | 2 - 3 | 270 | 62 | ||
| AK168012.1 | - | 2 - 3 | 274 | 62 | ||
| AK168230.1 | - | 2 - 3 | 269 | 62 | ||
| AK169034.1 | - | 2 - 3 | 274 | 62 | ||
| BC050927.1 | - | 1 - 2 | 333 | 6415084 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hspa5
Genomic Map