Assay Details
Gene Symbol: Pip5kl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198191.2 | NP_937834.1 | 1 - 2 | 73 | 78 | |
GenBank mRNA | AY376879.1 | - | 1 - 2 | 112 | 78 | |
BC094346.1 | - | 1 - 2 | 174 | 6411828 | 78 | |
BC117018.1 | - | 1 - 2 | 65 | 40126337 | 78 | |
BC119037.1 | - | 1 - 2 | 65 | 8733786 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
