Assay Details
| Gene Symbol: Pter | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012076.1 | - | 3 - 4 | 824 | 73 | |
| BC003793.1 | - | 3 - 4 | 791 | 3601675 | 73 | |
| U28016.1 | - | 3 - 4 | 800 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pter
Genomic Map