Assay Details
| Gene Symbol: Vim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011701.4 | NP_035831.2 | 7 - 8 | 1387 | 62 | |
| GenBank mRNA | AK012469.1 | - | 7 - 8 | 890 | 62 | |
| AK076001.1 | - | 7 - 8 | 1384 | 62 | ||
| AK133726.1 | - | 7 - 8 | 1388 | 62 | ||
| AK150270.1 | - | 7 - 8 | 1083 | 62 | ||
| AK151196.1 | - | 7 - 8 | 1390 | 62 | ||
| AK153019.1 | - | 7 - 8 | 1390 | 62 | ||
| AK159539.1 | - | 7 - 8 | 1388 | 62 | ||
| AK169181.1 | - | 7 - 8 | 1389 | 62 | ||
| AK170429.1 | - | 7 - 8 | 1388 | 62 | ||
| AK203121.1 | - | 2 - 3 | 56 | 62 | ||
| AK203726.1 | - | 3 - 4 | 309 | 62 | ||
| AK205443.1 | - | 2 - 3 | 177 | 62 | ||
| AK213831.1 | - | 3 - 4 | 315 | 62 | ||
| BC089335.1 | - | 7 - 8 | 1383 | 6415629 | 62 | |
| M24849.1 | - | 7 - 8 | 1304 | 62 | ||
| M26251.1 | - | 7 - 8 | 1746 | 62 | ||
| X51438.1 | - | 7 - 8 | 1356 | 62 | ||
| X56397.1 | - | 7 - 8 | 1350 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vim
Genomic Map