Assay Details
| Gene Symbol: Tor2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152800.3 | NP_690013.1 | 3 - 4 | 658 | 69 | |
| NR_045485.1 | 3 - 4 | 572 | 69 | |||
| GenBank mRNA | AK162719.1 | - | 3 - 4 | 602 | 69 | |
| AK170979.1 | - | 2 - 3 | 1837 | 69 | ||
| AK171247.1 | - | 3 - 4 | 610 | 69 | ||
| BC003466.1 | - | 3 - 4 | 643 | 2649388 | 69 | |
| BC023085.1 | - | 1 - 2 | 157 | 69 | ||
| BC024469.1 | - | 3 - 4 | 621 | 4976022 | 69 | |
| CK031473.1 | - | 3 - 4 | 519 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tor2a
Genomic Map