Assay Details
| Gene Symbol: Tor2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152800.3 | NP_690013.1 | 4 - 5 | 786 | 69 | |
| NR_045485.1 | 4 - 5 | 700 | 69 | |||
| GenBank mRNA | AK162719.1 | - | 4 - 5 | 730 | 69 | |
| AK170979.1 | - | 3 - 4 | 1965 | 69 | ||
| AK171247.1 | - | 4 - 5 | 738 | 69 | ||
| BC003466.1 | - | 4 - 5 | 771 | 2649388 | 69 | |
| BC023085.1 | - | 2 - 3 | 285 | 69 | ||
| BC024469.1 | - | 4 - 5 | 749 | 4976022 | 69 | |
| CK031473.1 | - | 4 - 5 | 647 | 69 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21117290 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21117289 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21117285 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21117288 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21117286 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21117284 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88773 | BC024469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88773 | BC023085.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88773 | AK170979.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88773 | AK213575.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88773 | AK218716.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88773 | BC024469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88773 | BC023085.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88773 | AK170979.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88773 | AK213575.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88773 | AK218716.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411022 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580664 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676416 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 689247 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119888 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88773 | BC024469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88773 | BC023085.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88773 | AK170979.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88773 | AK213575.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88773 | AK218716.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88773 | BC024469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88773 | BC023085.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88773 | AK170979.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88773 | AK213575.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88773 | AK218716.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P314968 | BC024469.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P314968 | BC023085.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P314968 | AK170979.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P314968 | BC003466.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P314968 | NM_152800.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21117278 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119888 | - | - |
| Whole Mouse Genome G4122A | A_51_P314968 | BC024469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314968 | BC023085.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314968 | AK170979.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314968 | BC003466.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314968 | NM_152800.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88773 | BC024469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88773 | BC023085.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88773 | AK170979.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88773 | AK213575.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88773 | AK218716.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88773 | BC024469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88773 | BC023085.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88773 | AK170979.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88773 | AK213575.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88773 | AK218716.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111651 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119888 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01336570 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01280890 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01336571 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01280888 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01584149 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01280886 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21117286 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21117285 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21117288 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21117289 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21117284 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21117290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21117375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02007866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02091483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02102718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01280888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01336571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01982614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21117308 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10471523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10471521 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10471524 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10471520 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10471522 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423783_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438547_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10471519 | - | - |
| Mouse Genome 430 2.0 Array | 1423783_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438547_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1423783_at | - | - |
| Mouse Genome 430 2.0 Array | 1438547_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 30933_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 30933_at | - | - |
| Mouse Expression Set 430 | 1438547_PM_x_at | - | - |
| Mouse Expression Set 430 | 1423783_at | - | - |
| Mouse Expression Set 430 | 1423783_PM_at | - | - |
| Mouse Expression Set 430 | 1438547_x_at | - | - |
| Murine Genome U74v2 Set | 97557_PM_at | - | - |
| Murine Genome U74v2 Set | 97557_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423783_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438547_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423783_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438547_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423783_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423783_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438547_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438547_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1400026 | - | - |
| MouseWG-6 v2 Expression BeadChip | 102100204 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1400026 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102100204 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001400026 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2519489 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1241588 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2519489 | - | - |
| mouse-6 v1.1 (Illumina) | scl030933.5_3-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102100204 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1400026 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl030933.5_3-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102100204 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1400026 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2519488 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1241588 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2519488 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl030933.5_3-S | - | - |

Tor2a
Genomic Map