Assay Details
| Gene Symbol: Fpgs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010236.2 | NP_034366.2 | 1 - 2 | 185 | 120 | |
| GenBank mRNA | AK041952.1 | - | 1 - 2 | 145 | 120 | |
| AK157040.1 | - | 1 - 2 | 146 | 120 | ||
| BC005484.1 | - | 1 - 2 | 128 | 3485401 | 120 | |
| U32197.1 | - | 1 - 2 | 142 | 120 | ||
| U33557.1 | - | 1 - 2 | 162 | 120 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC005484.1 transcripts.
Set Membership: |
|

Fpgs
Genomic Map