Assay Details
| Gene Symbol: Dpp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK211757.1 | - | 3 - 4 | 272 | 72 | |
| BC052861.1 | - | 6 - 7 | 1595 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Dpp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK211757.1 | - | 3 - 4 | 272 | 72 | |
| BC052861.1 | - | 6 - 7 | 1595 | 72 | ||
Set Membership: |
|