Assay Details
| Gene Symbol: Sapcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081085.2 | NP_001074554.1 | 3 - 4 | 1177 | 72 | |
| NM_001290407.1 | NP_001277336.1 | 2 - 3 | 1075 | 72 | ||
| GenBank mRNA | AB197927.1 | - | 2 - 3 | 1075 | 72 | |
| AK008577.1 | - | 2 - 3 | 772 | 72 | ||
| AK154143.1 | - | 3 - 4 | 886 | 72 | ||
| AK163973.1 | - | 3 - 4 | 396 | 72 | ||
| AK164615.1 | - | 3 - 4 | 852 | 72 | ||
| AK166625.1 | - | 3 - 4 | 852 | 72 | ||
| BC070477.1 | - | 2 - 3 | 776 | 30656854 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006594 | NM_001081085.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P472217 | AK166625.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P472217 | BC070477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P472217 | AK154143.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P472217 | AK163973.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P472217 | AB197927.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P472217 | AK166625.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P472217 | BC070477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P472217 | AK154143.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P472217 | AK163973.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P472217 | AB197927.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4966 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333742 | AK049650.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333742 | AK136947.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333742 | BC053080.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333742 | BC059836.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333742 | NM_177681.3 | refseq_mrna_mouse_NM |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121821 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P472217 | AK166625.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P472217 | BC070477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P472217 | AK154143.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P472217 | AK163973.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P472217 | AB197927.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P472217 | AK166625.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P472217 | BC070477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P472217 | AK154143.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P472217 | AK163973.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P472217 | AB197927.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006594 | NM_001081085.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121821 | - | - |
| Whole Mouse Genome G4122A | A_51_P472217 | AK166625.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472217 | BC070477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472217 | AK154143.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472217 | AK163973.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472217 | AB197927.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472217 | AK166625.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472217 | BC070477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472217 | AK154143.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472217 | AK163973.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472217 | AB197927.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121821 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01173681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01173684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01173696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01369162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01473656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02181353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01627469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01279343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02181354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01473669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01627464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071741 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02266422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02266424 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10470008 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470013 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470009 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470004 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470003 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470006 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470007 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470005 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470012 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470010 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470002 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470011 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429405_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438615_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429404_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10470001 | - | - |
| Mouse Genome 430 2.0 Array | 1429405_at | - | - |
| Mouse Genome 430 2.0 Array | 1438615_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1429404_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429404_at | - | - |
| Mouse Genome 430 2.0 Array | 1438615_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1429405_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72080_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72080_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200002326.mm.1 | NM_001081085.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002326.mm.1 | TC0200000413.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200002329.mm.1 | NM_001081085.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002329.mm.1 | TC0200000413.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429404_at | - | - |
| Mouse Expression Set 430 | 1438615_PM_x_at | - | - |
| Mouse Expression Set 430 | 1429405_at | - | - |
| Mouse Expression Set 430 | 1429405_PM_at | - | - |
| Mouse Expression Set 430 | 1438615_x_at | - | - |
| Mouse Expression Set 430 | 1429404_PM_at | - | - |
| Murine Genome U74v2 Set | 169193_f_at | - | - |
| Murine Genome U74v2 Set | 169193_PM_f_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429404_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429405_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438615_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429405_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438615_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429404_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 713909 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2644800 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2644800 | - | - |

Sapcd2
Genomic Map