Assay Details
| Gene Symbol: Cacnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252533.1 | NP_001239462.1 | 2 - 3 | 337 | 57 | |
| NM_023116.4 | NP_075605.1 | 3 - 4 | 468 | 57 | ||
| NR_045533.1 | 2 - 3 | 412 | 57 | |||
| GenBank mRNA | AK020806.1 | - | 2 - 3 | 413 | 57 | |
| AK034054.1 | - | 3 - 4 | 469 | 57 | ||
| AK046048.1 | - | 2 - 3 | 325 | 57 | ||
| AK078220.1 | - | 2 - 3 | 193 | 57 | ||
| AK082315.1 | - | 3 - 4 | 352 | 57 | ||
| AK143332.1 | - | 1 - 2 | 144 | 57 | ||
| AM259383.2 | - | 2 - 3 | 337 | 57 | ||
| BC109156.1 | - | 3 - 4 | 400 | 40047524 | 57 | |
| BC109157.1 | - | 3 - 4 | 400 | 40047531 | 57 | |
| FM872406.1 | - | 2 - 3 | 293 | 57 | ||
| FM872407.1 | - | 2 - 3 | 399 | 57 | ||
| FM872408.1 | - | 3 - 4 | 467 | 57 | ||
| KJ789960.1 | - | 2 - 3 | 170 | 57 | ||
| L20343.1 | - | 2 - 3 | 247 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cacnb2
Genomic Map