Assay Details
| Gene Symbol: Cacnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252533.1 | NP_001239462.1 | 5 - 6 | 670 | 70 | |
| NM_023116.4 | NP_075605.1 | 6 - 7 | 801 | 70 | ||
| NR_045533.1 | 5 - 6 | 745 | 70 | |||
| GenBank mRNA | AK034054.1 | - | 6 - 7 | 802 | 70 | |
| AK078220.1 | - | 5 - 6 | 526 | 70 | ||
| AM259383.2 | - | 5 - 6 | 670 | 70 | ||
| BC109156.1 | - | 6 - 7 | 733 | 40047524 | 70 | |
| BC109157.1 | - | 6 - 7 | 733 | 40047531 | 70 | |
| FM872406.1 | - | 5 - 6 | 626 | 70 | ||
| FM872407.1 | - | 5 - 6 | 732 | 70 | ||
| FM872408.1 | - | 6 - 7 | 800 | 70 | ||
| L20343.1 | - | 5 - 6 | 580 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cacnb2
Genomic Map