Assay Details
| Gene Symbol: A230108P19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040333.1 | 1 - 2 | 66 | 79 | ||
| GenBank mRNA | AK039218.1 | - | 1 - 2 | 67 | 79 | |
| AK044786.1 | - | 1 - 2 | 67 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 10857 | Gm10857 |
AU015850, ENSMUSG00000075545 |
-52.7kb | Sense | Chr. 2 - 6132869 - 6140570 on NCBI Build GRCm38 |
| Downstream | predicted gene 13383 | Gm13383 | OTTMUSG00000011755 | 10.2kb | Anti-sense | Chr. 2 - 6256247 - 6263036 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1700002196.rn
Set Membership: |
|

A230108P19Rik
Genomic Map