Assay Details
| Gene Symbol: Brd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113574.1 | NP_001107046.1 | 8 - 9 | 1598 | 95 | |
| GenBank mRNA | AB206708.2 | - | 7 - 8 | 1409 | 95 | |
| AK037435.1 | - | 8 - 9 | 1580 | 95 | ||
| AK076472.1 | - | 8 - 9 | 1529 | 95 | ||
| AK076924.1 | - | 8 - 9 | 1678 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Brd3
Genomic Map