Assay Details
| Gene Symbol: 9230102O04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040511.1 | 2 - 3 | 410 | 69 | ||
| GenBank mRNA | AK020309.1 | - | 2 - 3 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 13219 | Gm13219 | OTTMUSG00000011007 | -416.3kb | Anti-sense | Chr. 2 - 9352413 - 9467458 on NCBI Build GRCm38 |
| Downstream | predicted gene 13262 | Gm13262 | OTTMUSG00000011195 | 18.5kb | Anti-sense | Chr. 2 - 9906165 - 9918096 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

9230102O04Rik
Genomic Map