Assay Details
| Gene Symbol: Nelfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021393.2 | NP_067368.2 | 7 - 8 | 1286 | 78 | |
| GenBank mRNA | AB041607.1 | - | 6 - 7 | 1407 | 78 | |
| AK080419.1 | - | 7 - 8 | 988 | 78 | ||
| AK087524.1 | - | 7 - 8 | 986 | 78 | ||
| AK173124.1 | - | 6 - 7 | 821 | 78 | ||
| AK189417.1 | - | 2 - 3 | 160 | 78 | ||
| AK214584.1 | - | 2 - 3 | 160 | 78 | ||
| BC004762.1 | - | 7 - 8 | 1174 | 3155904 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nelfb
Genomic Map