Assay Details
| Gene Symbol: Cel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009885.1 | NP_034015.1 | 6 - 7 | 772 | 82 | |
| GenBank mRNA | AK131818.1 | - | 6 - 7 | 790 | 82 | |
| AK131824.1 | - | 6 - 7 | 790 | 82 | ||
| BC006872.1 | - | 6 - 7 | 784 | 2646703 | 82 | |
| U33169.1 | - | 6 - 7 | 772 | 82 | ||
| U37386.1 | - | 6 - 7 | 776 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cel
Genomic Map