Assay Details
| Gene Symbol: Zak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023057.5 | NP_075544.1 | 17 - 18 | 1724 | 66 | |
| GenBank mRNA | AB049731.1 | - | 16 - 17 | 1507 | 66 | |
| AK132186.1 | - | 17 - 18 | 1725 | 66 | ||
| BC023718.1 | - | 17 - 18 | 1605 | 5346593 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zak
Genomic Map