Assay Details
| Gene Symbol: Zak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023057.5 | NP_075544.1 | 18 - 19 | 1797 | 78 | |
| GenBank mRNA | AB049731.1 | - | 17 - 18 | 1580 | 78 | |
| AK132186.1 | - | 18 - 19 | 1798 | 78 | ||
| BC023718.1 | - | 18 - 19 | 1678 | 5346593 | 78 | |
More Information
Set Membership: |
|

Zak
Genomic Map