Assay Details
| Gene Symbol: Zak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164791.1 | NP_001158263.1 | 6 - 7 | 631 | 72 | |
| NM_023057.5 | NP_075544.1 | 6 - 7 | 695 | 72 | ||
| NM_178084.4 | NP_835185.2 | 6 - 7 | 631 | 72 | ||
| GenBank mRNA | AB049731.1 | - | 5 - 6 | 478 | 72 | |
| AB049732.1 | - | 5 - 6 | 478 | 72 | ||
| AK045444.1 | - | 6 - 7 | 556 | 72 | ||
| AK087363.1 | - | 6 - 7 | 571 | 72 | ||
| AK132186.1 | - | 6 - 7 | 696 | 72 | ||
| AK162819.1 | - | 6 - 7 | 496 | 72 | ||
| BC023718.1 | - | 6 - 7 | 576 | 5346593 | 72 | |
More Information
Set Membership: |
|

Zak
Genomic Map