Assay Details
| Gene Symbol: Cd302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290660.1 | NP_001277589.1 | 5 - 6 | 851 | 72 | |
| GenBank mRNA | AK002812.1 | - | 5 - 6 | 492 | 72 | |
| BC005501.2 | - | 4 - 5 | 413 | 3489789 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cd302
Genomic Map