Assay Details
| Gene Symbol: Gpcpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042672.1 | NP_001036137.1 | 3 - 4 | 354 | 102 | |
| NM_001291050.1 | NP_001277979.1 | 3 - 4 | 351 | 102 | ||
| NM_001291052.1 | NP_001277981.1 | 3 - 4 | 351 | 102 | ||
| NM_027096.2 | NP_081372.1 | 3 - 4 | 351 | 102 | ||
| NM_028802.3 | NP_083078.3 | 3 - 4 | 351 | 102 | ||
| GenBank mRNA | AK009137.1 | - | 3 - 4 | 291 | 102 | |
| AK030645.1 | - | 3 - 4 | 305 | 102 | ||
| AK031658.1 | - | 3 - 4 | 354 | 102 | ||
| AK050318.1 | - | 3 - 4 | 265 | 102 | ||
| AK141374.1 | - | 3 - 4 | 240 | 102 | ||
| AK165952.1 | - | 3 - 4 | 316 | 102 | ||
| AK166454.1 | - | 3 - 4 | 316 | 102 | ||
| BC006887.1 | - | 3 - 4 | 275 | 102 | ||
| BC033408.1 | - | 3 - 4 | 281 | 5357683 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC033408.1, NM_001291050.1, NM_001291052.1, NM_027096.2 transcripts.
Set Membership: |
|

Gpcpd1
Genomic Map