Assay Details
| Gene Symbol: Gpcpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042672.1 | NP_001036137.1 | 9 - 10 | 1075 | 145 | |
| NM_001291050.1 | NP_001277979.1 | 8 - 9 | 948 | 145 | ||
| NM_001291051.1 | NP_001277980.1 | 8 - 9 | 978 | 145 | ||
| NM_001291052.1 | NP_001277981.1 | 8 - 9 | 996 | 145 | ||
| NM_027096.2 | NP_081372.1 | 7 - 8 | 872 | 145 | ||
| NM_028802.3 | NP_083078.3 | 9 - 10 | 1072 | 145 | ||
| GenBank mRNA | AK009563.1 | - | 2 - 3 | 291 | 145 | |
| AK030645.1 | - | 9 - 10 | 1026 | 145 | ||
| AK031658.1 | - | 8 - 9 | 951 | 145 | ||
| AK049491.1 | - | 1 - 2 | 161 | 145 | ||
| AK050318.1 | - | 9 - 10 | 987 | 145 | ||
| AK051728.1 | - | 1 - 2 | 135 | 145 | ||
| AK141374.1 | - | 9 - 10 | 962 | 145 | ||
| AK163419.1 | - | 8 - 9 | 918 | 145 | ||
| AK165952.1 | - | 9 - 10 | 1037 | 145 | ||
| AK166454.1 | - | 8 - 9 | 961 | 145 | ||
| BC006887.1 | - | 8 - 9 | 920 | 145 | ||
| BC033408.1 | - | 7 - 8 | 802 | 5357683 | 145 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049491.1, AK051728.1 transcripts.
Set Membership: |
|

Gpcpd1
Genomic Map