Assay Details
| Gene Symbol: Ssfa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080558.4 | NP_542125.3 | 11 - 12 | 2889 | 91 | |
| GenBank mRNA | AB093303.1 | - | 11 - 12 | 2764 | 91 | |
| AB120565.1 | - | 11 - 12 | 2774 | 91 | ||
| AK037213.1 | - | 9 - 10 | 2599 | 91 | ||
| BC008560.1 | - | 4 - 5 | 1862 | 91 | ||
| BC018526.1 | - | 1 - 2 | 93 | 91 | ||
| BC079535.1 | - | 5 - 6 | 2268 | 91 | ||
| BC122519.1 | - | 11 - 12 | 2775 | 91 | ||
| BC141883.1 | - | 11 - 12 | 2774 | 40097831 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037213.1, BC008560.1 transcripts.
Set Membership: |
|

Ssfa2
Genomic Map