Assay Details
| Gene Symbol: Ssfa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080558.4 | NP_542125.3 | 5 - 6 | 514 | 108 | |
| GenBank mRNA | AB093303.1 | - | 5 - 6 | 389 | 108 | |
| AB120565.1 | - | 5 - 6 | 399 | 108 | ||
| AK037213.1 | - | 3 - 4 | 224 | 108 | ||
| AK169543.1 | - | 5 - 6 | 515 | 108 | ||
| BC122519.1 | - | 5 - 6 | 400 | 108 | ||
| BC141883.1 | - | 5 - 6 | 399 | 40097831 | 108 | |
| CF539296.1 | - | 5 - 6 | 330 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037213.1 transcripts.
Set Membership: |
|

Ssfa2
Genomic Map