Assay Details
| Gene Symbol: Ssfa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080558.4 | NP_542125.3 | 7 - 8 | 820 | 69 | |
| GenBank mRNA | AB093303.1 | - | 7 - 8 | 695 | 69 | |
| AB120565.1 | - | 7 - 8 | 705 | 69 | ||
| AK037213.1 | - | 5 - 6 | 530 | 69 | ||
| AK169543.1 | - | 7 - 8 | 821 | 69 | ||
| BC079535.1 | - | 1 - 2 | 199 | 69 | ||
| BC122519.1 | - | 7 - 8 | 706 | 69 | ||
| BC141883.1 | - | 7 - 8 | 705 | 40097831 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037213.1 transcripts.
Set Membership: |
|

Ssfa2
Genomic Map