Assay Details
| Gene Symbol: Ssfa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080558.4 | NP_542125.3 | 8 - 9 | 1530 | 131 | |
| GenBank mRNA | AB093303.1 | - | 8 - 9 | 1405 | 131 | |
| AB120565.1 | - | 8 - 9 | 1415 | 131 | ||
| AK037213.1 | - | 6 - 7 | 1240 | 131 | ||
| AK169543.1 | - | 8 - 9 | 1531 | 131 | ||
| BC008560.1 | - | 1 - 2 | 503 | 131 | ||
| BC079535.1 | - | 2 - 3 | 909 | 131 | ||
| BC122519.1 | - | 8 - 9 | 1416 | 131 | ||
| BC141883.1 | - | 8 - 9 | 1415 | 40097831 | 131 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037213.1, BC008560.1 transcripts.
Set Membership: |
|

Ssfa2
Genomic Map