Assay Details
| Gene Symbol: Fbxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212433.1 | NP_997598.1 | 10 - 11 | 1301 | 136 | |
| GenBank mRNA | AK004544.2 | - | 10 - 11 | 1253 | 136 | |
| AK009024.1 | - | 10 - 11 | 1240 | 136 | ||
| AK019550.1 | - | 10 - 11 | 1241 | 136 | ||
| AK028048.1 | - | 10 - 11 | 1263 | 136 | ||
| AK029594.1 | - | 3 - 4 | 420 | 136 | ||
| AK031185.1 | - | 10 - 11 | 1238 | 136 | ||
| AK031428.1 | - | 10 - 11 | 1261 | 136 | ||
| AK049870.1 | - | 10 - 11 | 1259 | 136 | ||
| AK050195.1 | - | 10 - 11 | 1253 | 136 | ||
| AK151130.1 | - | 10 - 11 | 1252 | 136 | ||
| AK151857.1 | - | 10 - 11 | 1252 | 136 | ||
| AK152780.1 | - | 10 - 11 | 1252 | 136 | ||
| AK167075.1 | - | 10 - 11 | 1239 | 136 | ||
| AK176959.1 | - | 3 - 4 | 336 | 136 | ||
| BC010212.1 | - | 10 - 11 | 1202 | 136 | ||
| BC050758.1 | - | 10 - 11 | 1254 | 136 | ||
| BC058253.1 | - | 10 - 11 | 1258 | 6465107 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo3
Genomic Map