Assay Details
| Gene Symbol: Mettl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029790.3 | NP_084066.2 | 1 - 2 | 100 | 125 | |
| GenBank mRNA | AK002682.1 | - | 1 - 2 | 101 | 125 | |
| BC089533.1 | - | 1 - 2 | 90 | 125 | ||
| BC106186.1 | - | 1 - 2 | 72 | 30244889 | 125 | |
| BC115926.1 | - | 1 - 2 | 59 | 40105262 | 125 | |
| BC117549.1 | - | 1 - 2 | 59 | 40105259 | 125 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002682.1, BC106186.1, BC115926.1, BC117549.1, NM_029790.3 transcripts.
Set Membership: |
|

Mettl15
Genomic Map