Assay Details
| Gene Symbol: Pax6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244201.1 | NP_001231130.1 | 4 - 5 | 421 | 65 | |
| NM_001244202.1 | NP_001231131.1 | 4 - 5 | 977 | 65 | ||
| GenBank mRNA | AF443223.1 | - | 1 - 2 | 136 | 65 | |
| AK028059.1 | - | 4 - 5 | 422 | 65 | ||
| AK045805.1 | - | 4 - 5 | 978 | 65 | ||
| BC036957.1 | - | 4 - 5 | 417 | 4504106 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pax6
Genomic Map