Assay Details
| Gene Symbol: Pax6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244202.1 | NP_001231131.1 | 1 - 2 | 712 | 86 | |
| NM_013627.5 | NP_038655.1 | 1 - 2 | 712 | 86 | ||
| GenBank mRNA | BC011272.1 | - | 1 - 2 | 400 | 4008490 | 86 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC011272.1, NM_001244202.1, NM_013627.5 transcripts.
Set Membership: |
|

Pax6
Genomic Map