Assay Details
| Gene Symbol: Ehf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007914.3 | NP_031940.1 | 1 - 2 | 142 | 80 | |
| GenBank mRNA | AK028651.1 | - | 1 - 2 | 113 | 80 | |
| BC005520.1 | - | 1 - 2 | 104 | 3494644 | 80 | |
| BC006789.1 | - | 1 - 2 | 124 | 3589528 | 80 | |
| BE134913.1 | - | 1 - 2 | 142 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC005520.1, BC006789.1, NM_007914.3 transcripts.
Set Membership: |
|

Ehf
Genomic Map