Assay Details
| Gene Symbol: Ehf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007914.3 | NP_031940.1 | 4 - 5 | 545 | 139 | |
| GenBank mRNA | AF035527.1 | - | 3 - 4 | 543 | 139 | |
| AK028651.1 | - | 4 - 5 | 516 | 139 | ||
| BC005520.1 | - | 5 - 6 | 462 | 3494644 | 139 | |
| BC006789.1 | - | 4 - 5 | 527 | 3589528 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ehf
Genomic Map