Assay Details
| Gene Symbol: Ehf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007914.3 | NP_031940.1 | 6 - 7 | 687 | 79 | |
| GenBank mRNA | AF035527.1 | - | 5 - 6 | 685 | 79 | |
| AK028651.1 | - | 6 - 7 | 658 | 79 | ||
| BC005520.1 | - | 7 - 8 | 604 | 3494644 | 79 | |
| BC006789.1 | - | 6 - 7 | 669 | 3589528 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ehf
Genomic Map