Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200008273.mm.1 |
NM_001165980.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008273.mm.1 |
NM_001165980.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008273.mm.1 |
NM_001165981.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008273.mm.1 |
NM_001165981.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008273.mm.1 |
NM_198005.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008273.mm.1 |
NM_198005.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008273.mm.1 |
TC0200001030.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008272.mm.1 |
NM_001165980.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008272.mm.1 |
NM_001165980.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008272.mm.1 |
NM_001165981.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008272.mm.1 |
NM_001165981.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008272.mm.1 |
NM_198005.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008272.mm.1 |
NM_198005.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008272.mm.1 |
TC0200001030.mm.1 |
MTA TC |