Assay Details
| Gene Symbol: Fbxl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178674.4 | NP_848789.2 | 2 - 3 | 209 | 65 | |
| GenBank mRNA | AK046196.1 | - | 2 - 3 | 205 | 65 | |
| AK085598.1 | - | 2 - 3 | 210 | 65 | ||
| BC138940.1 | - | 2 - 3 | 187 | 8861962 | 65 | |
| BC138941.1 | - | 2 - 3 | 187 | 8861963 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK085598.1, BC138940.1, BC138941.1, NM_178674.4 transcripts.
Set Membership: |
|

Fbxl21
Genomic Map