Assay Details
| Gene Symbol: Fbxl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178674.4 | NP_848789.2 | 3 - 4 | 340 | 98 | |
| GenBank mRNA | AK046196.1 | - | 3 - 4 | 336 | 98 | |
| AK085598.1 | - | 3 - 4 | 341 | 98 | ||
| BC138940.1 | - | 3 - 4 | 318 | 8861962 | 98 | |
| BC138941.1 | - | 3 - 4 | 318 | 8861963 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK085598.1, BC138940.1, BC138941.1, NM_178674.4 transcripts.
Set Membership: |
|

Fbxl21
Genomic Map