Assay Details
| Gene Symbol: Tmem164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199360.1 | NP_001186289.1 | 3 - 4 | 918 | 94 | |
| NM_177592.4 | NP_808260.2 | 3 - 4 | 861 | 94 | ||
| GenBank mRNA | AK089342.1 | - | 3 - 4 | 768 | 94 | |
| AK165839.1 | - | 3 - 4 | 754 | 94 | ||
| AK165910.1 | - | 3 - 4 | 754 | 94 | ||
| BC056470.1 | - | 3 - 4 | 843 | 5706838 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem164
Genomic Map