Assay Details
| Gene Symbol: Agps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172666.3 | NP_766254.2 | 8 - 9 | 946 | 70 | |
| GenBank mRNA | AK031049.1 | - | 8 - 9 | 948 | 70 | |
| AK031154.1 | - | 8 - 9 | 787 | 70 | ||
| AK042745.1 | - | 8 - 9 | 786 | 70 | ||
| AK053686.1 | - | 8 - 9 | 935 | 70 | ||
| AK079447.1 | - | 7 - 8 | 626 | 70 | ||
| BC063086.1 | - | 8 - 9 | 931 | 6332574 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agps
Genomic Map