Assay Details
| Gene Symbol: Muc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172979.3 | NP_766567.1 | 3 - 4 | 1084 | 66 | |
| GenBank mRNA | AK028832.1 | - | 2 - 3 | 279 | 66 | |
| AK052840.1 | - | 3 - 4 | 971 | 66 | ||
| AK142590.1 | - | 3 - 4 | 971 | 66 | ||
| AY198336.1 | - | 3 - 4 | 1002 | 66 | ||
| AY198337.1 | - | 3 - 4 | 969 | 66 | ||
| BC055469.1 | - | 2 - 3 | 252 | 66 | ||
| BC116796.1 | - | 3 - 4 | 963 | 40126115 | 66 | |
| BC119176.1 | - | 3 - 4 | 963 | 8733925 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971834 | BC119176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971834 | BC055469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971834 | BC094506.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971834 | BC116796.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971834 | BC020027.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138591 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162844 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971834 | BC119176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971834 | BC055469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971834 | BC094506.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971834 | BC116796.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971834 | BC020027.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21522033 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04645518 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138591 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04645493 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162844 | - | - |
| Whole Mouse Genome G4122A | A_51_P406807 | BC055469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406807 | BC094506.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406807 | BC020027.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406807 | BC038066.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406807 | BC048853.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P342627 | BC119176.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P342627 | BC055469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P342627 | BC094506.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P342627 | BC116796.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P342627 | BC020027.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105195 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138591 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162844 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01157273 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01157271 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02029584 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01157255 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01157261 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01157265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21522111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01693579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01568396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01486219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01344951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01922672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02199608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522017 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01157255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01220011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01652341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01415411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01486210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21522109 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10474471 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10474473 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10474474 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10474475 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10474472 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10474470 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10474468 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426941_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10474467 | - | - |
| Mouse Genome 430 2.0 Array | 1426941_at | - | - |
| Mouse Genome 430 2.0 Array | 1426941_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200012248.mm.1 | NM_172979.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200012248.mm.1 | TC0200001637.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200012249.mm.1 | NM_172979.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200012249.mm.1 | TC0200001637.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200012251.mm.1 | NM_172979.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200012251.mm.1 | TC0200001637.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200012250.mm.1 | NM_172979.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200012250.mm.1 | TC0200001637.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426941_PM_at | - | - |
| Mouse Expression Set 430 | 1426941_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426941_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426941_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426941_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426941_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101190452 | - | - |
| MouseWG-6 v2 Expression BeadChip | 2360541 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101190452 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2360541 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002360541 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2669326 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2669326 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2451987 | - | - |
| mouse-6 v1.1 (Illumina) | scl00269328.2_53-S | - | - |
| mouse-6 v1.1 (Illumina) | scl20498.5_293-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2360541 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101190452 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00269328.2_53-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2360541 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101190452 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2669326 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2451987 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2669326 | - | - |

Muc15
Genomic Map