Assay Details
| Gene Symbol: Qser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123327.2 | NP_001116799.1 | 5 - 6 | 4327 | 149 | |
| GenBank mRNA | AK141733.1 | - | 3 - 4 | 2638 | 149 | |
| CF749097.1 | - | 2 - 3 | 237 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Qser1
Genomic Map