Assay Details
| Gene Symbol: Qser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123327.2 | NP_001116799.1 | 6 - 7 | 4458 | 150 | |
| GenBank mRNA | AK141733.1 | - | 4 - 5 | 2769 | 150 | |
| CF749097.1 | - | 3 - 4 | 368 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Qser1
Genomic Map